Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCTGGGGATGTTGAGGAGCTTC[A/G]CTGCCACCAGGGGGTTGCTGTAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137217 MIM: 180381 MIM: 189902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP4B - ATPase H+/K+ transporting beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000705.3 | 797 | Missense Mutation | GCG,GTG | A,V 248 | NP_000696.1 |
GRK1 - G protein-coupled receptor kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002929.2 | 797 | Intron | NP_002920.1 | |||
XM_017020684.1 | 797 | Intron | XP_016876173.1 |
TFDP1 - transcription factor Dp-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |