Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAAGGTTAGTCCAGTTGAGAGC[A/G]CTTGATTATCTGGACTGAGAATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606205 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC6A7 - solute carrier family 6 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014228.4 | Intron | NP_055043.2 | ||||
XM_017009767.1 | Intron | XP_016865256.1 | ||||
XM_017009768.1 | Intron | XP_016865257.1 | ||||
XM_017009769.1 | Intron | XP_016865258.1 | ||||
XM_017009770.1 | Intron | XP_016865259.1 |