Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAAAACGATGAGGCTTCTTATG[G/T]AGGGTTGCCACAACCCCTCTATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAXDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAXDC2 - fatty acid hydroxylase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032385.4 | 3959 | Intron | NP_115761.2 | |||
XM_005268358.2 | 3959 | Intron | XP_005268415.1 | |||
XM_006714753.2 | 3959 | Intron | XP_006714816.1 | |||
XM_017008965.1 | 3959 | UTR 3 | XP_016864454.1 | |||
XM_017008966.1 | 3959 | Intron | XP_016864455.1 |
LARP1 - La ribonucleoprotein domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015315.4 | 3959 | Intron | NP_056130.2 | |||
XM_005268404.3 | 3959 | Intron | XP_005268461.1 | |||
XM_005268406.3 | 3959 | Intron | XP_005268463.1 | |||
XM_005268407.3 | 3959 | Intron | XP_005268464.1 | |||
XM_005268408.3 | 3959 | Intron | XP_005268465.1 | |||
XM_006714775.2 | 3959 | Intron | XP_006714838.1 | |||
XM_006714776.3 | 3959 | Intron | XP_006714839.1 | |||
XM_011537614.1 | 3959 | Intron | XP_011535916.1 | |||
XM_011537615.1 | 3959 | Intron | XP_011535917.1 | |||
XM_011537616.1 | 3959 | Intron | XP_011535918.1 | |||
XM_011537617.1 | 3959 | Intron | XP_011535919.1 | |||
XM_011537618.2 | 3959 | Intron | XP_011535920.1 | |||
XM_017009283.1 | 3959 | Intron | XP_016864772.1 | |||
XM_017009284.1 | 3959 | Intron | XP_016864773.1 |