Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCCCGTCACCATGGTGGATGTC[A/G]ATGTCAATGTAGAGCACCCGAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605166 MIM: 611213 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC3 - histone deacetylase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003883.3 | 573 | Silent Mutation | ATC,ATT | I,I 169 | NP_003874.2 | |
XM_011537697.2 | 573 | UTR 5 | XP_011535999.1 |
RELL2 - RELT like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |