Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAATCCTAGGAACAACGGTGCT[A/G]ATAACATAAGGGCACAAGCACCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRELID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRELID2 - PRELI domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138492.5 | 1052 | UTR 3 | NP_612501.3 | |||
NM_182960.3 | 1052 | UTR 3 | NP_892005.1 | |||
NM_205846.2 | 1052 | UTR 3 | NP_995318.1 | |||
XM_017009127.1 | 1052 | Intron | XP_016864616.1 | |||
XM_017009128.1 | 1052 | Intron | XP_016864617.1 | |||
XM_017009129.1 | 1052 | Intron | XP_016864618.1 | |||
XM_017009130.1 | 1052 | Intron | XP_016864619.1 | |||
XM_017009131.1 | 1052 | Intron | XP_016864620.1 | |||
XM_017009132.1 | 1052 | Intron | XP_016864621.1 | |||
XM_017009133.1 | 1052 | Intron | XP_016864622.1 | |||
XM_017009134.1 | 1052 | Intron | XP_016864623.1 | |||
XM_017009135.1 | 1052 | Intron | XP_016864624.1 |