Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGAGCAAGGAACTGGGAGTACC[C/G]AAAGACCCTCTGCTCTTCATTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC4A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC4A9 - solute carrier family 4 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258426.1 | 188 | Silent Mutation | CCC,CCG | P,P 51 | NP_001245355.1 | |
NM_001258427.1 | 188 | Silent Mutation | CCC,CCG | P,P 51 | NP_001245356.1 | |
NM_001258428.1 | 188 | Silent Mutation | CCC,CCG | P,P 51 | NP_001245357.1 | |
NM_031467.2 | 188 | Silent Mutation | CCC,CCG | P,P 51 | NP_113655.2 | |
XM_005268521.3 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_005268578.2 | |
XM_011537694.2 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_011535996.1 | |
XM_017009933.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865422.1 | |
XM_017009934.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865423.1 | |
XM_017009935.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865424.1 | |
XM_017009936.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865425.1 | |
XM_017009937.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865426.1 | |
XM_017009938.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865427.1 | |
XM_017009939.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865428.1 | |
XM_017009940.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865429.1 | |
XM_017009941.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865430.1 | |
XM_017009942.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865431.1 | |
XM_017009943.1 | 188 | Silent Mutation | CCC,CCG | P,P 75 | XP_016865432.1 |