Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAGAACTCTTTCAAAAAAAAA[T/A]TGGGGATCAAAATACAAGTCTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606720 MIM: 611477 | ||||||||||||||||||||
Literature Links: |
ENDOU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ENDOU - endonuclease, poly(U) specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172439.1 | 2177 | UTR 3 | NP_001165910.1 | |||
NM_001172440.1 | 2177 | UTR 3 | NP_001165911.1 | |||
NM_006025.3 | 2177 | UTR 3 | NP_006016.1 |
LOC105369748 - uncharacterized LOC105369748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPAP3 - RNA polymerase II associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |