Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGTTGGGCCTCAACAAGCGAC[G/T]GGCTCTCCGCGGAAGCTTCAGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105369748 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105369748 - uncharacterized LOC105369748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAPGEF3 - Rap guanine nucleotide exchange factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098531.2 | 4026 | UTR 3 | NP_001092001.1 | |||
NM_001098532.2 | 4026 | UTR 3 | NP_001092002.1 | |||
NM_006105.5 | 4026 | UTR 3 | NP_006096.2 | |||
XM_005268571.3 | 4026 | Intron | XP_005268628.1 | |||
XM_011537752.1 | 4026 | Intron | XP_011536054.1 | |||
XM_011537755.1 | 4026 | Intron | XP_011536057.1 | |||
XM_011537758.2 | 4026 | Intron | XP_011536060.1 | |||
XM_017018688.1 | 4026 | Intron | XP_016874177.1 |