Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTGTGACACGCGAGGCTCCTC[A/G]GTTACTTAGCCAACGGCAGAGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600748 | ||||||||||||||||||||
Literature Links: |
TMBIM6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMBIM6 - transmembrane BAX inhibitor motif containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098576.1 | 363 | Missense Mutation | CAG,CGG | Q,R 12 | NP_001092046.1 | |
NM_003217.2 | 363 | Intron | NP_003208.2 | |||
XM_005269126.3 | 363 | Missense Mutation | CAG,CGG | Q,R 12 | XP_005269183.1 |