Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCCCAAGGGGCAGGAGGAATGC[A/G]GGAGCAGGAGTTGAGCTTGGGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142974 MIM: 142973 MIM: 142972 MIM: 142970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXC4 - homeobox C4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014620.5 | 294 | UTR 5 | NP_055435.2 | |||
NM_153633.2 | 294 | Intron | NP_705897.1 |
HOXC5 - homeobox C5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018953.3 | 294 | Intron | NP_061826.1 |
HOXC6 - homeobox C6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004503.3 | 294 | Intron | NP_004494.1 | |||
NM_153693.4 | 294 | UTR 5 | NP_710160.1 |
HOXC8 - homeobox C8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984512 - uncharacterized LOC107984512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |