Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGGCATAGGAGAAGAGGCTTT[A/C]AGAACAAGGGATACTCCTCACTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171891 MIM: 613246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE1B - phosphodiesterase 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R1A - protein phosphatase 1 regulatory inhibitor subunit 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006741.3 | Intron | NP_006732.3 | ||||
XM_005268995.4 | Intron | XP_005269052.1 | ||||
XM_006719471.3 | Intron | XP_006719534.1 |