Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGCTGTCACCTGATTATAAGAC[C/T]TCTTTTGCACCTTCCCAGGTGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601081 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCD2 - ATP binding cassette subfamily D member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005164.3 | 7912 | Intron | NP_005155.1 | |||
XM_011538027.2 | 7912 | UTR 3 | XP_011536329.1 | |||
XM_017018992.1 | 7912 | Intron | XP_016874481.1 | |||
XM_017018993.1 | 7912 | Intron | XP_016874482.1 | |||
XM_017018994.1 | 7912 | Intron | XP_016874483.1 |