Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAGGGCCCAGATAGAGGCCCAGG[A/T]AAGCAAATAAATGAAAGACGGTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616016 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPM1H - protein phosphatase, Mg2+/Mn2+ dependent 1H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020700.1 | 3489 | UTR 3 | NP_065751.1 | |||
XM_011538578.2 | 3489 | UTR 3 | XP_011536880.1 | |||
XM_011538579.1 | 3489 | UTR 3 | XP_011536881.1 | |||
XM_017019676.1 | 3489 | Intron | XP_016875165.1 |