Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGACTCCTATATGAGAGTCGC[A/G]AGTGTGCAGCAGGTAAAGGCATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609007 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRK2 - leucine rich repeat kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198578.3 | 462 | Silent Mutation | GCA,GCG | A,A 75 | NP_940980.3 | |
XM_005268629.3 | 462 | Silent Mutation | GCA,GCG | A,A 75 | XP_005268686.1 | |
XM_011537877.2 | 462 | Silent Mutation | GCA,GCG | A,A 75 | XP_011536179.1 | |
XM_011537881.2 | 462 | Silent Mutation | GCA,GCG | A,A 75 | XP_011536183.1 | |
XM_011537882.2 | 462 | Silent Mutation | GCA,GCG | A,A 75 | XP_011536184.1 | |
XM_017018786.1 | 462 | Silent Mutation | GCA,GCG | A,A 75 | XP_016874275.1 | |
XM_017018787.1 | 462 | Intron | XP_016874276.1 | |||
XM_017018788.1 | 462 | Intron | XP_016874277.1 | |||
XM_017018789.1 | 462 | Silent Mutation | GCA,GCG | A,A 75 | XP_016874278.1 |