Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTTCAAGAATCCTGACAGGCA[A/C]AATATCCACAAAAGAGCAAACAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616998 MIM: 616874 | ||||||||||||||||||||
Literature Links: |
LLPH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LLPH - LLP homolog, long-term synaptic facilitation | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LLPH-AS1 - LLPH antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMBIM4 - transmembrane BAX inhibitor motif containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282606.1 | 751 | Missense Mutation | TTG,TTT | L,F 210 | NP_001269535.1 | |
NM_001282609.1 | 751 | Intron | NP_001269538.1 | |||
NM_001282610.1 | 751 | Missense Mutation | TTG,TTT | L,F 132 | NP_001269539.1 | |
NM_016056.3 | 751 | Missense Mutation | TTG,TTT | L,F 163 | NP_057140.2 |