Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTTTTTCTGGCCAAGAACAGC[A/G]CAGCTCAAAGAGGTTAGCTGGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601871 MIM: 607799 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSRP2 - cysteine and glycine rich protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300965.1 | Intron | NP_001287894.1 | ||||
NM_001321.2 | Intron | NP_001312.1 |
ZDHHC17 - zinc finger DHHC-type containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |