Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACTTGCTGCTTTTTTTTTTTTT[A/T]TTTGGAAAGTTTGCTTATAACTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602021 | ||||||||||||||||||||
Literature Links: |
PPP1R12A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R12A - protein phosphatase 1 regulatory subunit 12A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005268887.3 | 3477 | UTR 3 | XP_005268944.1 | |||
XM_011538376.2 | 3477 | UTR 3 | XP_011536678.1 | |||
XM_011538378.2 | 3477 | UTR 3 | XP_011536680.1 | |||
XM_017019327.1 | 3477 | UTR 3 | XP_016874816.1 | |||
XM_017019330.1 | 3477 | Intron | XP_016874819.1 |