Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGTTAAAACATTAATTATCAGT[C/G]ACAAATGAAATCCAGAAACATAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEA1 - early endosome antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003566.3 | Intron | NP_003557.2 | ||||
XM_011538814.2 | Intron | XP_011537116.1 | ||||
XM_011538815.2 | Intron | XP_011537117.1 | ||||
XM_017020018.1 | Intron | XP_016875507.1 |
PLEKHG7 - pleckstrin homology and RhoGEF domain containing G7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |