Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACGTTCCCCATGCAACGTGAC[A/G]CCCTTTATTTTCATGGATTAGTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615847 MIM: 604259 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP83 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP83 - centrosomal protein 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042399.1 | Intron | NP_001035858.1 | ||||
NM_016122.2 | Intron | NP_057206.2 |
PLXNC1 - plexin C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005761.2 | Intron | NP_005752.1 | ||||
XM_006719186.3 | Intron | XP_006719249.1 | ||||
XM_011537730.2 | Intron | XP_011536032.1 | ||||
XM_011537731.2 | Intron | XP_011536033.1 | ||||
XM_017018671.1 | Intron | XP_016874160.1 | ||||
XM_017018672.1 | Intron | XP_016874161.1 |