Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTCTCCTGCCATGATATCTTGCT[C/G]ATGATATTCTGTGATCCTTTACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRAM1 - DNA damage regulated autophagy modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018370.2 | Intron | NP_060840.2 | ||||
XM_005269004.2 | Intron | XP_005269061.1 | ||||
XM_005269005.2 | Intron | XP_005269062.1 | ||||
XM_017019578.1 | Intron | XP_016875067.1 |