Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAATGTGGCTGGGCTTCATCCGC[G/A]GAGGGAGGAAAGAGGTGGCTCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600738 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SELPLG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SELPLG - selectin P ligand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206609.1 | 1675 | UTR 3 | NP_001193538.1 | |||
NM_003006.4 | 1675 | UTR 3 | NP_002997.2 |