Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGATTCTTAAGTACACTGAAGT[C/T]TGAGAACAGTTGATCTAAGTATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609539 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID2 - AT-rich interaction domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152641.2 | Intron | NP_689854.2 | ||||
XM_006719272.3 | Intron | XP_006719335.1 | ||||
XM_011538025.1 | Intron | XP_011536327.1 |
LINC00938 - long intergenic non-protein coding RNA 938 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |