Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATCAATCTTTGATTCTCCTTTG[A/C]AATGTTTTGATCACTTTTTTATGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMIGO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMIGO2 - adhesion molecule with Ig like domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143668.1 | 3717 | Intron | NP_001137140.1 | |||
NM_181847.4 | 3717 | UTR 3 | NP_862830.1 | |||
XM_005268837.3 | 3717 | Intron | XP_005268894.1 |
PCED1B - PC-esterase domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |