Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTGGGAGGAAGTAACCGGGAA[A/G]TCGCCACGGGGACACCCCTCACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMIGO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMIGO2 - adhesion molecule with Ig like domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143668.1 | 259 | Intron | NP_001137140.1 | |||
NM_181847.4 | 259 | Intron | NP_862830.1 | |||
XM_005268837.3 | 259 | UTR 5 | XP_005268894.1 |
PCED1B - PC-esterase domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281429.1 | 259 | Intron | NP_001268358.1 | |||
NM_138371.2 | 259 | Intron | NP_612380.1 | |||
XM_005269224.4 | 259 | Intron | XP_005269281.1 | |||
XM_011538978.2 | 259 | Intron | XP_011537280.1 | |||
XM_017020207.1 | 259 | Intron | XP_016875696.1 | |||
XM_017020208.1 | 259 | Intron | XP_016875697.1 | |||
XM_017020209.1 | 259 | Intron | XP_016875698.1 | |||
XM_017020210.1 | 259 | Intron | XP_016875699.1 | |||
XM_017020211.1 | 259 | Intron | XP_016875700.1 | |||
XM_017020212.1 | 259 | Intron | XP_016875701.1 | |||
XM_017020213.1 | 259 | Intron | XP_016875702.1 | |||
XM_017020214.1 | 259 | Intron | XP_016875703.1 | |||
XM_017020215.1 | 259 | Intron | XP_016875704.1 | |||
XM_017020216.1 | 259 | Intron | XP_016875705.1 |