Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAAGAATTAAATACCAAACATT[A/G]TTACATTCAACTCTGAAAGATGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM26 - transmembrane protein 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178505.7 | 4630 | UTR 3 | NP_848600.2 | |||
XM_011539450.2 | 4630 | Intron | XP_011537752.1 | |||
XM_011539451.1 | 4630 | Intron | XP_011537753.1 | |||
XM_017015829.1 | 4630 | Intron | XP_016871318.1 |