Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAATCTTTATTATTTCAAAATT[A/C]CTATGTTTACAGAGAATTTCTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604503 MIM: 609348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMJD1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JMJD1C - jumonji domain containing 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REEP3 - receptor accessory protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001330.2 | Intron | NP_001001330.1 | ||||
XM_011539501.2 | Intron | XP_011537803.1 | ||||
XM_017015896.1 | Intron | XP_016871385.1 |