Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTGCCCATAGTGCCCGACTACAT[C/G]GCCCACATGCGCGGGGGCGGCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118490 MIM: 600336 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHAT - choline O-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142929.1 | 623 | Intron | NP_001136401.1 | |||
NM_001142933.1 | 623 | Intron | NP_001136405.1 | |||
NM_001142934.1 | 623 | Intron | NP_001136406.1 | |||
NM_020549.4 | 623 | Intron | NP_065574.3 | |||
NM_020984.3 | 623 | Intron | NP_066264.3 | |||
NM_020985.3 | 623 | Intron | NP_066265.3 | |||
NM_020986.3 | 623 | Intron | NP_066266.3 |
SLC18A3 - solute carrier family 18 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003055.2 | 623 | Missense Mutation | ATC,ATG | I,M 61 | NP_003046.2 |