Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTAGTCAGGAAGGAAGATGTAAA[A/C]AAAAAAAAGAAAAACAGCCAACTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607790 | ||||||||||||||||||||
Literature Links: |
TET1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TET1 - tet methylcytosine dioxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030625.2 | 562 | Missense Mutation | AAA,AAC | K,N 19 | NP_085128.2 | |
XM_011540204.2 | 562 | Missense Mutation | AAA,AAC | K,N 19 | XP_011538506.1 | |
XM_011540205.2 | 562 | Intron | XP_011538507.1 | |||
XM_011540206.2 | 562 | Missense Mutation | AAA,AAC | K,N 19 | XP_011538508.1 | |
XM_011540207.2 | 562 | Intron | XP_011538509.1 | |||
XM_017016686.1 | 562 | Intron | XP_016872175.1 | |||
XM_017016687.1 | 562 | Intron | XP_016872176.1 | |||
XM_017016688.1 | 562 | Intron | XP_016872177.1 | |||
XM_017016689.1 | 562 | Intron | XP_016872178.1 |