Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTTACACAATACTACTCTTACT[G/A]AAATCCTAGAGCCAGCGAGTTAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605516 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf105 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf105 - chromosome 10 open reading frame 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164375.2 | 2518 | UTR 3 | NP_001157847.1 | |||
NM_001168390.1 | 2518 | UTR 3 | NP_001161862.1 | |||
XM_011539808.2 | 2518 | UTR 3 | XP_011538110.1 | |||
XM_011539809.2 | 2518 | UTR 3 | XP_011538111.1 |