Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTTGGACATTCTTTTCAAAGTA[C/T]TGAAATGGCCTGTACCACCACACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611573 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGMS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGMS1 - sphingomyelin synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147156.3 | 2079 | Silent Mutation | CAA,CAG | Q,Q 375 | NP_671512.1 | |
XM_005269675.1 | 2079 | Silent Mutation | CAA,CAG | Q,Q 375 | XP_005269732.1 | |
XM_011539582.2 | 2079 | Silent Mutation | CAA,CAG | Q,Q 375 | XP_011537884.1 | |
XM_011539583.2 | 2079 | Silent Mutation | CAA,CAG | Q,Q 375 | XP_011537885.1 | |
XM_011539584.2 | 2079 | Intron | XP_011537886.1 |