Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAACAAGAAGAGCAAAACAAAC[A/G]TATGTATTGGGTCCTTGCCATGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf99 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf99 - chromosome 10 open reading frame 99 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207373.2 | Intron | NP_997256.1 | ||||
XM_005269811.3 | Intron | XP_005269868.1 |
HOST2 - human ovarian cancer-specific transcript 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |