Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGAAAGGTTACCCCTTTCTAG[A/C]TCCACTATCATCTAATTTTATTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613922 MIM: 613924 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPK - lipase family member K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIPN - lipase family member N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102469.1 | Intron | NP_001095939.1 | ||||
XM_005270049.3 | Intron | XP_005270106.1 | ||||
XM_011540083.2 | Intron | XP_011538385.1 | ||||
XM_011540084.2 | Intron | XP_011538386.1 | ||||
XM_011540085.2 | Intron | XP_011538387.1 | ||||
XM_011540086.2 | Intron | XP_011538388.1 | ||||
XM_017016549.1 | Intron | XP_016872038.1 |