Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGTGCAGCTATTTCCTGCAAT[A/G]AGAGGAAGGCTTGGCAGGAGCGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFIT3 - interferon induced protein with tetratricopeptide repeats 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031683.3 | Intron | NP_001026853.1 | ||||
NM_001289758.1 | Intron | NP_001276687.1 | ||||
NM_001289759.1 | Intron | NP_001276688.1 | ||||
NM_001549.5 | Intron | NP_001540.2 |