Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATCTTTGGCTGTGAAGTGTTT[G/T]CCTACCCCATGGCCTCCATCGAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609208 | ||||||||||||||||||||
Literature Links: |
KAZALD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KAZALD1 - Kazal type serine peptidase inhibitor domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319303.1 | 1514 | Missense Mutation | GCC,TCC | A,S 59 | NP_001306232.1 | |
NM_030929.4 | 1514 | Missense Mutation | GCC,TCC | A,S 197 | NP_112191.2 | |
XM_005270194.3 | 1514 | Missense Mutation | GCC,TCC | A,S 197 | XP_005270251.1 | |
XM_017016714.1 | 1514 | Missense Mutation | GCC,TCC | A,S 197 | XP_016872203.1 | |
XM_017016715.1 | 1514 | Missense Mutation | GCC,TCC | A,S 197 | XP_016872204.1 |