Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGATGAATAGATAGATTTTGACA[A/G]ATATCTCTCTGAGGTAGGCAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTRC - beta-transducin repeat containing E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256856.1 | Intron | NP_001243785.1 | ||||
NM_003939.4 | Intron | NP_003930.1 | ||||
NM_033637.3 | Intron | NP_378663.1 | ||||
XM_006718054.2 | Intron | XP_006718117.1 | ||||
XM_011540320.2 | Intron | XP_011538622.1 | ||||
XM_017016870.1 | Intron | XP_016872359.1 | ||||
XM_017016871.1 | Intron | XP_016872360.1 | ||||
XM_017016872.1 | Intron | XP_016872361.1 | ||||
XM_017016873.1 | Intron | XP_016872362.1 | ||||
XM_017016874.1 | Intron | XP_016872363.1 |