Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGAACACAAATGTGTAAAAAT[A/G]AGAGTGTTCATTAAAGCATTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613605 MIM: 608610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBIP1 - BBSome interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195304.1 | Intron | NP_001182233.1 | ||||
NM_001195305.1 | Intron | NP_001182234.1 | ||||
NM_001195306.1 | Intron | NP_001182235.1 | ||||
NM_001195307.1 | Intron | NP_001182236.1 | ||||
NM_001243783.1 | Intron | NP_001230712.1 |
MIR4680 - microRNA 4680 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDCD4 - programmed cell death 4 (neoplastic transformation inhibitor) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |