Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCATTCTCCTCACCATATATTCA[A/G]AGGGCAGAAGGCCCTCTAAATCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC186 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC186 - coiled-coil domain containing 186 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321829.1 | 4789 | UTR 3 | NP_001308758.1 | |||
NM_018017.3 | 4789 | UTR 3 | NP_060487.2 | |||
NM_153249.1 | 4789 | Intron | NP_694981.1 | |||
XM_011539915.2 | 4789 | UTR 3 | XP_011538217.1 | |||
XM_011539916.2 | 4789 | Intron | XP_011538218.1 |