Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATATTTGGCTAGTCAGGAAGACA[A/G]TTCAGTTTCTTCAAACCCGAATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605796 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TDRD1 - tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198795.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | NP_942090.1 | |
XM_005269978.2 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_005270035.1 | |
XM_005269979.2 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_005270036.1 | |
XM_011539959.2 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538261.1 | |
XM_011539960.2 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538262.1 | |
XM_011539961.2 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538263.1 | |
XM_011539962.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538264.1 | |
XM_011539963.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538265.1 | |
XM_011539964.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538266.1 | |
XM_011539966.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_011538268.1 | |
XM_017016414.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_016871903.1 | |
XM_017016415.1 | 415 | Missense Mutation | AAT,AGT | N,S 83 | XP_016871904.1 |