Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCAAAATCTGAGATGCCTGTAA[C/T]ATATTGCATTATAGTGATCCGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606417 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR11 - WD repeat domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018117.11 | Intron | NP_060587.8 | ||||
XM_005269962.2 | Intron | XP_005270019.1 | ||||
XM_005269963.2 | Intron | XP_005270020.1 | ||||
XM_005269964.2 | Intron | XP_005270021.1 | ||||
XM_017016397.1 | Intron | XP_016871886.1 | ||||
XM_017016398.1 | Intron | XP_016871887.1 | ||||
XM_017016399.1 | Intron | XP_016871888.1 | ||||
XM_017016400.1 | Intron | XP_016871889.1 |
WDR11-AS1 - WDR11 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |