Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATAGCACTTCTGATTAATACTC[C/T]GGCTGTTTATTTTTAAAGAAATAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM175B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM175B - family with sequence similarity 175 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032182.3 | Intron | NP_115558.3 | ||||
XM_011539554.1 | Intron | XP_011537856.1 | ||||
XM_017015986.1 | Intron | XP_016871475.1 |