Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAGTTTTATTTCTCTCTGGGTA[C/T]CTTTTTCAGATCTCCAGGCCAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGBD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGBD2 - piggyBac transposable element derived 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017434.1 | Intron | NP_001017434.1 | ||||
NM_170725.2 | Intron | NP_733843.1 | ||||
XM_005270333.1 | Intron | XP_005270390.1 | ||||
XM_011544159.2 | Intron | XP_011542461.1 | ||||
XM_011544161.2 | Intron | XP_011542463.1 |