Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCAAAAGACTTTATACATGTCA[A/G]GTACCCTCTGCTTCCTGGTTCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF672 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF672 - zinc finger protein 672 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF692 - zinc finger protein 692 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136036.2 | Intron | NP_001129508.1 | ||||
NM_001193328.1 | Intron | NP_001180257.1 | ||||
NM_017865.3 | Intron | NP_060335.2 | ||||
XM_011544220.1 | Intron | XP_011542522.1 | ||||
XM_011544221.1 | Intron | XP_011542523.1 | ||||
XM_011544222.1 | Intron | XP_011542524.1 | ||||
XM_011544223.2 | Intron | XP_011542525.1 | ||||
XM_011544227.1 | Intron | XP_011542529.1 | ||||
XM_017001698.1 | Intron | XP_016857187.1 | ||||
XM_017001699.1 | Intron | XP_016857188.1 | ||||
XM_017001700.1 | Intron | XP_016857189.1 | ||||
XM_017001701.1 | Intron | XP_016857190.1 | ||||
XM_017001702.1 | Intron | XP_016857191.1 | ||||
XM_017001703.1 | Intron | XP_016857192.1 | ||||
XM_017001704.1 | Intron | XP_016857193.1 | ||||
XM_017001705.1 | Intron | XP_016857194.1 |