Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCTTGGCTGAACTGATCAAGTC[A/G]ATAGTTGCTTGACAAACTCACCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601476 MIM: 115437 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAPTM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAPTM5 - lysosomal protein transmembrane 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006762.2 | 1031 | UTR 3 | NP_006753.1 | |||
XM_011542098.2 | 1031 | UTR 3 | XP_011540400.1 |
MATN1 - matrilin 1, cartilage matrix protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MATN1-AS1 - MATN1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4420 - microRNA 4420 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |