Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTGTTGTCCTCCAACTGCAAT[A/C]AGGAAAAGGAAGGACACAGTTGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC17 - coiled-coil domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPBP1L1 - GC-rich promoter binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021639.4 | Intron | NP_067652.1 | ||||
XM_017002019.1 | Intron | XP_016857508.1 |