Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCCTTCCATTTCTGAATCTGTA[A/G]AATGGGGATAACATTTTCTACCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMBX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMBX1 - diencephalon/mesencephalon homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147192.2 | Intron | NP_671725.1 | ||||
NM_172225.1 | Intron | NP_757379.1 | ||||
XM_011540668.2 | Intron | XP_011538970.1 | ||||
XM_017000289.1 | Intron | XP_016855778.1 |