Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAGAAGGCCTCATTTCTGTTAA[C/T]TTATTGCCACCTACTGGCCTACGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4X1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP4X1 - cytochrome P450 family 4 subfamily X member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320289.1 | Intron | NP_001307218.1 | ||||
NM_001320290.1 | Intron | NP_001307219.1 | ||||
NM_178033.1 | Intron | NP_828847.1 | ||||
XM_017000973.1 | Intron | XP_016856462.1 |