Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAAACCCGCCAAGGGCCACCCT[A/T]GGGGAACTCGAGGGATTGAGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZYG11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZYG11A - zyg-11 family member A, cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004339.2 | Intron | NP_001004339.2 | ||||
NM_001307931.1 | Intron | NP_001294860.1 | ||||
XM_011541480.2 | Intron | XP_011539782.1 | ||||
XM_011541481.2 | Intron | XP_011539783.1 | ||||
XM_011541482.2 | Intron | XP_011539784.1 | ||||
XM_011541483.2 | Intron | XP_011539785.1 | ||||
XM_011541484.1 | Intron | XP_011539786.1 | ||||
XM_011541485.1 | Intron | XP_011539787.1 | ||||
XM_011541486.1 | Intron | XP_011539788.1 | ||||
XM_011541487.2 | Intron | XP_011539789.1 | ||||
XM_011541488.1 | Intron | XP_011539790.1 |