Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGGCCATAGGAGGAGTGACTAA[C/T]TTATTCTTTAGTCAAACAGCACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM59 - transmembrane protein 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305043.1 | 2205 | UTR 3 | NP_001291972.1 | |||
NM_001305049.1 | 2205 | UTR 3 | NP_001291978.1 | |||
NM_001305050.1 | 2205 | UTR 3 | NP_001291979.1 | |||
NM_001305051.1 | 2205 | UTR 3 | NP_001291980.1 | |||
NM_001305052.1 | 2205 | UTR 3 | NP_001291981.1 | |||
NM_001305066.1 | 2205 | Intron | NP_001291995.1 | |||
NM_004872.4 | 2205 | UTR 3 | NP_004863.2 | |||
XM_006711051.1 | 2205 | Intron | XP_006711114.1 |