Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCGCGCGGGTGTCGCAACATCC[C/G]CATTCACAATTCCCGACCAAGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC22 - tetratricopeptide repeat domain 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114108.1 | 1957 | UTR 3 | NP_001107580.1 | |||
NM_017904.3 | 1957 | Intron | NP_060374.2 | |||
XM_011541671.2 | 1957 | UTR 3 | XP_011539973.1 | |||
XM_017001582.1 | 1957 | UTR 3 | XP_016857071.1 |