Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCTTCTAGTGGCTTAAAAATG[C/T]TTTCCTTGAGAAATACACGATCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607786 MIM: 610569 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCSK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCSK9 - proprotein convertase subtilisin/kexin type 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP24 - ubiquitin specific peptidase 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015306.2 | 8332 | UTR 3 | NP_056121.2 | |||
XM_005270690.3 | 8332 | UTR 3 | XP_005270747.1 | |||
XM_017000831.1 | 8332 | UTR 3 | XP_016856320.1 | |||
XM_017000832.1 | 8332 | UTR 3 | XP_016856321.1 | |||
XM_017000833.1 | 8332 | UTR 3 | XP_016856322.1 | |||
XM_017000834.1 | 8332 | UTR 3 | XP_016856323.1 | |||
XM_017000835.1 | 8332 | Intron | XP_016856324.1 | |||
XM_017000836.1 | 8332 | Intron | XP_016856325.1 | |||
XM_017000837.1 | 8332 | Intron | XP_016856326.1 |